Niobium foil, 2.0mm (0.08in) thick, 99.8% (metals basis)
Niobium foil, 2.0mm (0.08in) thick, 99.8% (metals basis)
Niobium foil, 2.0mm (0.08in) thick, 99.8% (metals basis)
Thermo Scientific Chemicals

Niobium foil, 2.0mm (0.08in) thick, 99.8% (metals basis)

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 100 mm
100 x 200 mm
Catalog number 043231.FY
also known as 043231-FY
Price (USD)
375.65
Online exclusive
417.00
Save 41.35
Each
Quantity:
50 x 100 mm
Request bulk or custom format
Price (USD)
375.65
Online exclusive
417.00
Save 41.35
Each
Specifications
Assay Percent Notes(metals basis)
CAS3-1-7440
ChEBICHEBI:33344
Chemical Name or MaterialNiobium foil
EINECS Number231-113-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only