Tin foil, 2.0mm (0.08in) thick, 99.8% (metals basis)
Tin foil, 2.0mm (0.08in) thick, 99.8% (metals basis)
Tin foil, 2.0mm (0.08in) thick, 99.8% (metals basis)
Thermo Scientific Chemicals

Tin foil, 2.0mm (0.08in) thick, 99.8% (metals basis)

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Catalog number 043236.FI
also known as 043236-FI
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.80
Save 9.15
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.80
Save 9.15
Each
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.8% (metals basis)
FormFoil, 2.0mm (0.08in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only