2-Amino-4-methylbenzonitrile, 98%
2-Amino-4-methylbenzonitrile, 98%
2-Amino-4-methylbenzonitrile, 98%
Thermo Scientific Chemicals

2-Amino-4-methylbenzonitrile, 98%

CAS: 26830-96-6 | C8H8N2 | 132.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number 043260.14
also known as 043260-14
Price (USD)
635.65
Special offer
Online exclusive
Ends: 30-Jun-2026
748.00
Save 112.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
635.65
Special offer
Online exclusive
Ends: 30-Jun-2026
748.00
Save 112.35 (15%)
Each
Specifications
CAS26830-96-6
Chemical Name or Material2-Amino-4-methylbenzonitrile
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECS Number248-020-0
Formula Weight132.17
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only