Indium wire, 2.0mm (0.08in) dia, 99.99% (metals basis)
Indium wire, 2.0mm (0.08in) dia, 99.99% (metals basis)
Indium wire, 2.0mm (0.08in) dia, 99.99% (metals basis)
Thermo Scientific™

Indium wire, 2.0mm (0.08in) dia, 99.99% (metals basis)

Form: Approx.22.95g/m | CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
1 m
5 m
Catalog number 043265.BU
also known as 043265-BU
Price (USD)
76.65
Special offer
Online exclusive
Ends: 28-Dec-2026
90.00
Save 13.35
Each
Quantity:
25 cm
Request bulk or custom format
Price (USD)
76.65
Special offer
Online exclusive
Ends: 28-Dec-2026
90.00
Save 13.35
Each
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.01%
FormWire, 2.0mm dia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only