Silver(I) oxide, Electrical Grade
Silver(I) oxide, Electrical Grade
Silver(I) oxide, Electrical Grade
Thermo Scientific Chemicals

Silver(I) oxide, Electrical Grade

For battery plates, pollution control filters | CAS: 20667-12-3 | Ag2O | 231.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 043268.30
also known as 043268-30
Price (USD)
1,149.00
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,149.00
Each
Chemical Identifiers
CAS20667-12-3
IUPAC Name(argentiooxy)silver
Molecular FormulaAg2O
InChI KeyKHJDQHIZCZTCAE-UHFFFAOYSA-N
SMILES[Ag]O[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ag: ≥ 90%
Assay (unspecified)Silver Oxide: ≥97.5%
Particle Size-325 mesh
Trace MetalReport metal impurities analysis
FormPowder
For battery plates, and pollution control filters. It is used in the preparation of other silver compounds, and silver-oxide batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only