Platinum 20 wt% Rhodium wire, 0.25mm (0.01in) dia, 99.9% (metals basis)
Thermo Scientific Chemicals

Platinum 20 wt% Rhodium wire, 0.25mm (0.01in) dia, 99.9% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 043274.BU
also known as 043274-BU
Price (USD)
-
Quantity:
25 cm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangePt:Rh; 80:20 wt%
Chemical Name or MaterialPlatinum 20 wt% Rhodium wire
MDL NumberMFCD00198186
Name Note0.25mm (0.01 in.) dia.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only