Brilliant Blue G, ultrapure
Brilliant Blue G, ultrapure
Brilliant Blue G, ultrapure
Thermo Scientific Chemicals

Brilliant Blue G, ultrapure

Protein staining following gel electrophoresis | CAS: 6104-58-1 | C47H48N3NaO7S2 | 854.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 043318.18
also known as 043318-18
Price (USD)
168.65
Special offer
Online exclusive
Ends: 30-Jun-2026
198.00
Save 29.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
168.65
Special offer
Online exclusive
Ends: 30-Jun-2026
198.00
Save 29.35 (15%)
Each
Chemical Identifiers
CAS6104-58-1
IUPAC Namesodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}-2-methylphenyl)methylium
Molecular FormulaC47H48N3NaO7S2
InChI KeyCMGWKYXJIYYESN-UHFFFAOYSA-M
SMILES[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
CommentC.I. 42655
Loss on Drying≤10% (105?C, 3h)
Assay (Dye)≥65%
CommentSuitable for protein staining
View more
Brilliant Blue G is used in analytical biochemistry dye and as a P2X7 antagonist. It is also used in protein staining following gel electrophoresis. It is used to prepare the protein reagent for the determination of protein content of the collagenase enzyme isolated from fish waste. It may be employed as stain for the internal limiting membrane (ILM) for the macular hole (MH) and epiretinal membrane (ERM) surgery. It is useful for SDS gels as it readily stains proteins with minimal background color. Protein bands can be visualized during staining. It has been used in the Bradford dye-binding protein assay. A mechanism for dye binding to protein has been proposed, based on measuring Coomassie Brilliant Blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids. Brilliant Blue G has been used for determining critical micelle concentration of detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only