Platinum Iridium wire, 0.127mm (0.005in) dia, hard, 99.9% (metals basis excluding precious metals)
Thermo Scientific Chemicals

Platinum Iridium wire, 0.127mm (0.005in) dia, hard, 99.9% (metals basis excluding precious metals)

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
50 cm
Catalog number 043330.BW
also known as 043330-BW
Price (USD)
482.00
Each
Quantity:
50 cm
Request bulk or custom format
Price (USD)
482.00
Each
Specifications
Assay Percent Notes(metals basis excluding precious metals)
Avg. Mol. Wt. or Mol. Wt. RangePt:Ir; 80:20 wt%
Chemical Name or MaterialPlatinum Iridium wire
MDL NumberMFCD02091683
Name Note0.127mm (0.005 in.) dia., Hard
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only