Tin rod, 12.7mm (0.5in) dia, 99.98% (metals basis)
Tin rod, 12.7mm (0.5in) dia, 99.98% (metals basis)
Tin rod, 12.7mm (0.5in) dia, 99.98% (metals basis)
Thermo Scientific Chemicals

Tin rod, 12.7mm (0.5in) dia, 99.98% (metals basis)

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 043341.BU
also known as 043341-BU
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Quantity:
25 cm
Request bulk or custom format
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-31-5
ChEBICHEBI:32990
Chemical Name or MaterialTin rod
Density7.310 g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only