1,3-Diethylbenzene, 97+%
1,3-Diethylbenzene, 97+%
1,3-Diethylbenzene, 97+%
Thermo Scientific Chemicals

1,3-Diethylbenzene, 97+%

CAS: 141-93-5 | C10H14 | 134.222 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 mL
10 mL
Catalog number 043369.S0
also known as 043369-S0
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.50
Save 6.85 (14%)
Each
Quantity:
2 mL
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.50
Save 6.85 (14%)
Each
Specifications
Assay Percent Range>97%
CAS141-93-5
Chemical Name or Material1,3-Diethylbenzene
ColorColorless
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE
View more
1,3-Diethylbenzene is used as an intermediate and solvent. It is also used in the preparation of divinyl benzene. Further, it is used in powderless etching.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only