5-Hexyn-1-ol, 97%
5-Hexyn-1-ol, 97%
5-Hexyn-1-ol, 97%
Thermo Scientific Chemicals

5-Hexyn-1-ol, 97%

CAS: 928-90-5 | C6H10O | 98.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 043370.03
also known as 043370-03
Price (USD)
41.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
41.10
Each
Chemical Identifiers
CAS928-90-5
IUPAC Namehex-5-yn-1-ol
Molecular FormulaC6H10O
InChI KeyGOQJMMHTSOQIEI-UHFFFAOYSA-N
SMILESOCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 96.0%
5-Hexyn-1-ol is used as starting material for the preparation of terminal alkynes and lactones containing terminal alykyne groups. It is also used in the synthesis of cinnoline-fused cyclic enediyne, llycopodium alkaloids, (+)-nankakurine A and (+)-nankakurine B and 7-benzoyloxy-3-(2-nitrophenylseleno)-1,5-cyclodecadiyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only