2-Bromo-5-(trifluoromethyl)pyridine, 96%
2-Bromo-5-(trifluoromethyl)pyridine, 96%
2-Bromo-5-(trifluoromethyl)pyridine, 96%
Thermo Scientific Chemicals

2-Bromo-5-(trifluoromethyl)pyridine, 96%

CAS: 50488-42-1 | C6H3BrF3N | 225.996 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043381.03
also known as 043381-03
Price (USD)
64.65
Online Exclusive
72.00
Save 7.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
64.65
Online Exclusive
72.00
Save 7.35 (10%)
Each
Chemical Identifiers
CAS50488-42-1
IUPAC Name2-bromo-5-(trifluoromethyl)pyridine
Molecular FormulaC6H3BrF3N
InChI KeyGSKMWMFOQQBVMI-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(Br)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)White to pale cream
Melting Point (clear melt)44-47?C
Assay (GC)≥95.0%
2-Bromo-5-(trifluoromethyl)pyridine is a substrate used in a palladium-catalyzed α-arylation of a Refomatsky reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only