Indium rod, 2.38mm (0.094in) dia, 99.99% (metals basis)
Indium rod, 2.38mm (0.094in) dia, 99.99% (metals basis)
Indium rod, 2.38mm (0.094in) dia, 99.99% (metals basis)
Thermo Scientific Chemicals

Indium rod, 2.38mm (0.094in) dia, 99.99% (metals basis)

CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 cm
5 x 20 cm
Catalog number 043388.RQ
also known as 043388-RQ
Price (USD)
91.65
Special offer
Online exclusive
Ends: 28-Dec-2026
108.00
Save 16.35
Each
Quantity:
20 cm
Request bulk or custom format
Price (USD)
91.65
Special offer
Online exclusive
Ends: 28-Dec-2026
108.00
Save 16.35
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-74-6
ChEBICHEBI:30430
Chemical Name or MaterialIndium rod
EINECS Number231-180-0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only