Glassy carbon spherical powder, 10-20 micron, type 1
Glassy carbon spherical powder, 10-20 micron, type 1
Thermo Scientific Chemicals

Glassy carbon spherical powder, 10-20 micron, type 1

Widely used as an electrode material in electrochemistry. It is used to make carbon paste electrodes in electrochemistry.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 043489.18
also known as 043489-18
Price (USD)
212.00
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
212.00
Each
Add to cart
Specifications
Chemical Name or MaterialGlassy carbon spherical powder
DOT InformationHazard Class: 4.1; Packaging Group: III
Health Hazard 1H228
Health Hazard 3P210-P240-P241-P280-P370+P378q
MDL NumberMFCD00133992
View more
Glassy carbon spherical powder is widely used as an electrode material in electrochemistry. It is used to make carbon paste electrodes in electrochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.