Glassy carbon spherical powder, 10-20 micron, type 2
Thermo Scientific Chemicals

Glassy carbon spherical powder, 10-20 micron, type 2

Used as a catalyst support for bimetallic Rh-ReOx. It is also used to enhance the accuracy of infrared (IR) surface temperature measurements on micro-electro-mechanical systems (MEMS).
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 043490.30
also known as 043490-30
Price (USD)
907.00
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
907.00
Each
Specifications
Chemical Name or MaterialGlassy carbon spherical powder
EINECS Number231-153-3
MDL NumberMFCD00133992
PackagingGlass bottle
Recommended StorageAmbient temperatures
View more
Glassy carbon is used as a catalyst support for bimetallic Rh-ReOx. It is also used to enhance the accuracy of infrared (IR) surface temperature measurements on micro-electro-mechanical systems (MEMS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.