Scandium rod, 12.7mm (0.5in) dia, 99.6% (metals basis excluding Ta)
Scandium rod, 12.7mm (0.5in) dia, 99.6% (metals basis excluding Ta)
Scandium rod, 12.7mm (0.5in) dia, 99.6% (metals basis excluding Ta)
Thermo Scientific Chemicals

Scandium rod, 12.7mm (0.5in) dia, 99.6% (metals basis excluding Ta)

CAS: 7440-20-2 | Sc | 44.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
25 mm
Catalog number 043499.DH
also known as 043499-DH
Price (USD)
1,435.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,689.00
Save 253.35
Each
Quantity:
25 mm
Request bulk or custom format
Price (USD)
1,435.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,689.00
Save 253.35
Each
Chemical Identifiers
CAS7440-20-2
IUPAC Namescandium
Molecular FormulaSc
InChI KeySIXSYDAISGFNSX-UHFFFAOYSA-N
SMILES[Sc]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Ta0.4% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged in mylar under argon
RUO – Research Use Only