Sodium chloride, Spectroscopy Grade
Sodium chloride, Spectroscopy Grade
Sodium chloride, Spectroscopy Grade
Thermo Scientific Chemicals

Sodium chloride, Spectroscopy Grade

CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Catalog NumberQuantity
043631.19
also known as 043631-19
75 g
Catalog number 043631.19
also known as 043631-19
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Quantity:
75 g
Request bulk or custom format
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Specifications
Boiling Point1413°C
CAS7647-14-5
ChEBICHEBI:26710
Chemical Name or MaterialSodium chloride
ColorWhite
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only