Calcium iodide, ultra dry, 99.99% (metals basis)
Calcium iodide, ultra dry, 99.99% (metals basis)
Calcium iodide, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Calcium iodide, ultra dry, 99.99% (metals basis)

Light Sensitive and Hygroscopic | CAS: 10102-68-8 | CaI2 | 293.887 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 043674.14
also known as 043674-14
Price (USD)
144.65
Special offer
Online exclusive
Ends: 28-Dec-2026
170.00
Save 25.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
144.65
Special offer
Online exclusive
Ends: 28-Dec-2026
170.00
Save 25.35
Each
Chemical Identifiers
CAS10102-68-8
IUPAC Namecalcium diiodide
Molecular FormulaCaI2
InChI KeyUNMYWSMUMWPJLR-UHFFFAOYSA-L
SMILES[Ca++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤0.01%
Appearance (Color)White
Residual water≤100ppm
Calcium iodide is used in photography. It is also used in cat food as a source of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only