Titanium foil, 0.5mm (0.02in) thick, stress relieved, 99.99+% (metals basis)
Titanium foil, 0.5mm (0.02in) thick, stress relieved, 99.99+% (metals basis)
Titanium foil, 0.5mm (0.02in) thick, stress relieved, 99.99+% (metals basis)
Thermo Scientific Chemicals

Titanium foil, 0.5mm (0.02in) thick, stress relieved, 99.99+% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 043676.GH
also known as 043676-GH
Price (USD)
902.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,062.00
Save 159.35 (15%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
902.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,062.00
Save 159.35 (15%)
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.5mm (0.02in) thick, stress relieved.
Assay from Supplier's CofA>99.99% (metals basis)
Appearance (Color)Silvery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only