Polyethylene oxide, M.W. 1,000,000
Polyethylene oxide, M.W. 1,000,000
Polyethylene oxide, M.W. 1,000,000
Polyethylene oxide, M.W. 1,000,000
Thermo Scientific Chemicals

Polyethylene oxide, M.W. 1,000,000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 043678.22
also known as 043678-22
Price (USD)
166.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
166.00
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Viscosity400 to 800 cps ( 2% aq. solution, 25 deg C)
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only