Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles
Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles
Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles
Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles

CAS: 111-69-3 | C6H8N2 | 108.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 043679.AP
also known as 043679-AP
Price (USD)
87.65
Online Exclusive
97.00
Save 9.35 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
87.65
Online Exclusive
97.00
Save 9.35 (10%)
Each
Chemical Identifiers
CAS111-69-3
IUPAC Namehexanedinitrile
Molecular FormulaC6H8N2
InChI KeyBTGRAWJCKBQKAO-UHFFFAOYSA-N
SMILESN#CCCCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear, colorless to pale yellow
Assay from Supplier's CofA≥99.0%
Infrared spectrumConforms
Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only