Iodomethane, 99+%, stab. with copper
Iodomethane, 99+%, stab. with copper
Iodomethane, 99+%, stab. with copper
Thermo Scientific Chemicals

Iodomethane, 99+%, stab. with copper

CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 043681.18
also known as 043681-18
Price (USD)
36.20
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
36.20
Each
Chemical Identifiers
CAS74-88-4
IUPAC Nameiodomethane
Molecular FormulaCH3I
InChI KeyINQOMBQAUSQDDS-UHFFFAOYSA-N
SMILESCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (special)≥99.0%
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the in situ formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only