Calcium molybdenum oxide, 99%
Calcium molybdenum oxide, 99%
Calcium molybdenum oxide, 99%
Thermo Scientific Chemicals

Calcium molybdenum oxide, 99%

CAS: 7789-82-4 | CaMoO4 | 200.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 043686.14
also known as 043686-14
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.50
Save 6.85
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.50
Save 6.85
Each
Chemical Identifiers
CAS7789-82-4
IUPAC Namecalcium dioxomolybdenumbis(olate)
Molecular FormulaCaMoO4
InChI KeyBIOOACNPATUQFW-UHFFFAOYSA-N
SMILES[Ca++].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only