Palladium(II) sulfate dihydrate, Premion™, 99.995% (metals basis)
Palladium(II) sulfate dihydrate, Premion™, 99.995% (metals basis)
Palladium(II) sulfate dihydrate, Premion™, 99.995% (metals basis)
Thermo Scientific Chemicals

Palladium(II) sulfate dihydrate, Premion™, 99.995% (metals basis)

CAS: 13444-98-9 | H4O6PdS | 238.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043698.03
also known as 043698-03
Price (USD)
156.65
Special offer
Online exclusive
Ends: 28-Dec-2026
184.00
Save 27.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
156.65
Special offer
Online exclusive
Ends: 28-Dec-2026
184.00
Save 27.35
Each
Chemical Identifiers
CAS13444-98-9
IUPAC Namepalladium(2+) dihydrate sulfate
Molecular FormulaH4O6PdS
InChI KeyTWIRRPLUAGEFNJ-UHFFFAOYSA-L
SMILESO.O.[Pd++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only