Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min
Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min
Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min
Thermo Scientific Chemicals

Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min

Used in electroplating, as a catalyst and analytical reagent. Also used in the production of semiconductor epitaxy, diffusion process. | CAS: 13454-96-1 | Cl4Pt | 336.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043703.06
also known as 043703-06
Price (USD)
822.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
822.00
Each
Chemical Identifiers
CAS13454-96-1
IUPAC Nametetrachloroplatinum
Molecular FormulaCl4Pt
InChI KeyFBEIPJNQGITEBL-UHFFFAOYSA-J
SMILESCl[Pt](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pt 57% min
Total Metal Impurities0.01% max.
Platinum(IV) chloride is used in electroplating, as a catalyst and analytical reagent. Also used in the production of semiconductor epitaxy, diffusion process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only