2,3-Dimethylpentane, 97%
2,3-Dimethylpentane, 97%
2,3-Dimethylpentane, 97%
2,3-Dimethylpentane, 97%
Thermo Scientific Chemicals

2,3-Dimethylpentane, 97%

CAS: 565-59-3 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 043707.06
also known as 043707-06
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.10
Save 9.45 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.10
Save 9.45 (15%)
Each
Add to cart
Specifications
CAS565-59-3
Chemical Name or Material2,3-Dimethylpentane
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
Density0.690 g/mL
EINECS Number209-280-0
View more
It is used in the liquid-phase oxidation of 2,4-dimethylpentane and in a study on the preparation and the physical constants of a number of alkanes and cycloalkanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only