Polystyrene latex microsphere, 1 micron, dry form
Polystyrene latex microsphere, 1 micron, dry form
Polystyrene latex microsphere, 1 micron, dry form
Thermo Scientific Chemicals

Polystyrene latex microsphere, 1 micron, dry form

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
043711.ME500 mg
Catalog number 043711.ME
Price (USD)
625.65
Online Exclusive
695.00
Save 69.35 (10%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
625.65
Online Exclusive
695.00
Save 69.35 (10%)
Each
Specifications
CAS9003-53-6
ChEBICHEBI:27452
Chemical Name or MaterialPolystyrene latex microsphere
IUPAC Namestyrene
InChI KeyPPBRXRYQALVLMV-UHFFFAOYSA-N
View more
Polystyrene is used in adhesive, plastics, coatings, for electrical applications. In laboratories containers such as tubes, petridishes were made in polystyrene, used for the building construction panel packing, insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only