Polystyrene latex microsphere, carboxylated, 0.35 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, carboxylated, 0.35 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, carboxylated, 0.35 micron, 2.5 wt% dispersion in water
Thermo Scientific Chemicals

Polystyrene latex microsphere, carboxylated, 0.35 micron, 2.5 wt% dispersion in water

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
043717.4Y
also known as 043717-4Y
15 mL
Catalog number 043717.4Y
also known as 043717-4Y
Price (USD)
456.65
Online Exclusive
507.00
Save 50.35 (10%)
Each
Quantity:
15 mL
Request bulk or custom format
Price (USD)
456.65
Online Exclusive
507.00
Save 50.35 (10%)
Each
Specifications
CAS9003-53-6
ChEBICHEBI:27452
Chemical Name or MaterialPolystyrene latex microsphere
IUPAC Nameethenylbenzene
InChI KeyPPBRXRYQALVLMV-UHFFFAOYSA-N
View more
These particles are widely used in phagocytosis and neural retrograde transport studies, and as markers for cell bound antigens. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only