Cesium chloride, ultra dry, 99.99% (metals basis)
Cesium chloride, ultra dry, 99.99% (metals basis)
Cesium chloride, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Cesium chloride, ultra dry, 99.99% (metals basis)

CAS: 7647-17-8 | ClCs | 168.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 043720.09
also known as 043720-09
Price (USD)
138.65
Online Exclusive
154.00
Save 15.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
138.65
Online Exclusive
154.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS7647-17-8
IUPAC Namecaesium(1+) chloride
Molecular FormulaClCs
InChI KeyAIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES[Cl-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBeads
Residual water≤100 ppm
Total Metal Impurities≤0.01%
Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only