3-Hydroxy-3-methyl-2-butanone, 90+%
3-Hydroxy-3-methyl-2-butanone, 90+%
3-Hydroxy-3-methyl-2-butanone, 90+%
Thermo Scientific Chemicals

3-Hydroxy-3-methyl-2-butanone, 90+%

CAS: 115-22-0 | C5H10O2 | 102.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 043724.06
also known as 043724-06
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.90
Save 7.25 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.90
Save 7.25 (15%)
Each
Specifications
CAS115-22-0
Chemical Name or Material3-Hydroxy-3-methyl-2-butanone
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Density0.96g/mL
Flash Point42°C (107.6°F)
View more
3-Hydroxy-3-methyl-2-butanone used to produce 2,5-dihydroxy-2-methyl-heptan-3-one, laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only