Tantalum(V) chloride, anhydrous, Puratronic™, 99.995% (metals basis)
Tantalum(V) chloride, anhydrous, Puratronic™, 99.995% (metals basis)
Tantalum(V) chloride, anhydrous, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Tantalum(V) chloride, anhydrous, Puratronic™, 99.995% (metals basis)

CAS: 7721-01-9 | Cl5Ta | 358.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 043731.06
also known as 043731-06
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS7721-01-9
IUPAC Nametantalum(5+) pentachloride
Molecular FormulaCl5Ta
InChI KeyOEIMLTQPLAGXMX-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only