Leucomalachite Green
Leucomalachite Green
Leucomalachite Green
Thermo Scientific Chemicals

Leucomalachite Green

CAS: 129-73-7 | C23H26N2 | 330.475 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 043758.09
also known as 043758-09
Price (USD)
103.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
103.00
Each
Chemical Identifiers
CAS129-73-7
IUPAC Name4-{[4-(dimethylamino)phenyl](phenyl)methyl}-N,N-dimethylaniline
Molecular FormulaC23H26N2
InChI KeyWZKXBGJNNCGHIC-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale green or pale blue
FormCrystals or powder or crystalline powder
CommentMaterial sourced in US and in other countries
Elemental Analysis (typical)Carbon, C: 77.74-89.45% [US from vendor's CofA]
Elemental Analysis (typical)Nitrogen, N: 7.88-9.07% [US from vendor's CofA]
View more
Leucomalachite Green is a reagent for the quantitative colorimetric micro determination of hemoglobin and other heme compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only