Trimethyl(pentamethylcyclopentadienyl)titanium(IV), 97%
Trimethyl(pentamethylcyclopentadienyl)titanium(IV), 97%
Trimethyl(pentamethylcyclopentadienyl)titanium(IV), 97%
Thermo Scientific Chemicals

Trimethyl(pentamethylcyclopentadienyl)titanium(IV), 97%

CAS: 107333-47-1 | C13H29Ti-5 | 233.242 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 043759.02
also known as 043759-02
Price (USD)
180.65
Special offer
Online exclusive
Ends: 30-Jun-2026
212.00
Save 31.35 (15%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
180.65
Special offer
Online exclusive
Ends: 30-Jun-2026
212.00
Save 31.35 (15%)
Each
Specifications
CAS107333-47-1
Chemical Name or MaterialTrimethyl (pentamethylcyclopentadienyl)titanium (IV)
DOT InformationTransport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Formula Weight228.22
Health Hazard 1H228-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only