Specifications
CAS107333-47-1
Chemical Name or MaterialTrimethyl (pentamethylcyclopentadienyl)titanium (IV)
DOT InformationTransport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Formula Weight228.22
Health Hazard 1H228-H319-H335
View more