Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%
Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%
Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%
Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%
Thermo Scientific Chemicals

Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%

CAS: 16570-28-8 | Cu2O7P2 | 301.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 043760.A1
also known as 043760-A1
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.40
Save 12.75 (15%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.40
Save 12.75 (15%)
Each
Chemical Identifiers
CAS16570-28-8
IUPAC Namedicopper(2+) (phosphonatooxy)phosphonate
Molecular FormulaCu2O7P2
InChI KeyPEVJCYPAFCUXEZ-UHFFFAOYSA-J
SMILES[Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Cu 33.5-35.5%
Copper(II) pyrophosphate hydrate is used in printed circuit boards. It catalyzes the dimerization of isobutylene to diisobutylene. It is involved in the electrodeposition of copper-zinc alloys based electrolytes. Further, it is used in animal feed as a supplement. In addition to this, it is used in plating of copper on plastics, aluminum, and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only