Trimethyl borate, 99.99% (metals basis)
Trimethyl borate, 99.99% (metals basis)
Trimethyl borate, 99.99% (metals basis)
Thermo Scientific Chemicals

Trimethyl borate, 99.99% (metals basis)

CAS: 121-43-7 | C3H9BO3 | 103.912 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 043765.14
also known as 043765-14
Price (USD)
43.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
43.00
Each
Specifications
Assay Percent Notes(metals basis)
Beilstein1697939
CAS121-43-7
ChEBICHEBI:38913
Chemical Name or MaterialTrimethyl borate
View more
Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in the Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only