2-Chloro-2-methylnonane, 90+%
2-Chloro-2-methylnonane, 90+%
2-Chloro-2-methylnonane, 90+%
Thermo Scientific Chemicals

2-Chloro-2-methylnonane, 90+%

CAS: 4325-50-2 | C10H21Cl | 176.728 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 043784.14
also known as 043784-14
Price (USD)
284.65
Special offer
Online exclusive
Ends: 30-Jun-2026
335.00
Save 50.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
284.65
Special offer
Online exclusive
Ends: 30-Jun-2026
335.00
Save 50.35 (15%)
Each
Specifications
CAS4325-50-2
Chemical Name or Material2-Chloro-2-methylnonane
ColorColorless
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Formula Weight176.73
View more
It is used in the photoelectron spectroscopy of C60Fn- and C60Fm2 in the gas phase and the generation and characterization of C1-C60F47- and D2-C60F44 in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only