Pyridine, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles
Pyridine, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles
Pyridine, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Pyridine, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles

CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 043799.K2
also known as 043799-K2
Price (USD)
173.65
Special offer
Online exclusive
Ends: 30-Jun-2026
204.00
Save 30.35 (15%)
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
173.65
Special offer
Online exclusive
Ends: 30-Jun-2026
204.00
Save 30.35 (15%)
Each
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Color (APHA)≤10
Assay (GC)≥99.5%
FormLiquid
Water Content (Karl Fischer Titration)≤0.05% (≤500 ppm)
View more
Pyridine is used in spectrophotometry and environmental testing.As polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further, it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane and pyridine-sulfur trioxide are widely used owing to their superior characteristics in their reactivity and solubility characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only