Cyanide, standard solution, Specpure™ CN∣- 1000μg/mL
Thermo Scientific Chemicals

Cyanide, standard solution, Specpure™ CN∣- 1000μg/mL

KCN in 0.1% NaOH
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 043804.AE
also known as 043804-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialCyanide
ColorColorless
Concentration or Composition (by Analyte or Components)CN: 1000μg/mL
Density1g/cm3 at 20°C
Health Hazard 1H301-H312
View more
Cyanide, standard solution is used to dissolve metals and their ores. It is also used in clinical chemistry and in waste water treatment facilities to determine the concentration of dissolved and particulate materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only