Copper plate, irregular, nominal 10x34x0.6cm (4x13.5x0.25in), Puratronic™, 99.999% (metals basis)
Copper plate, irregular, nominal 10x34x0.6cm (4x13.5x0.25in), Puratronic™, 99.999% (metals basis)
Copper plate, irregular, nominal 10x34x0.6cm (4x13.5x0.25in), Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Copper plate, irregular, nominal 10x34x0.6cm (4x13.5x0.25in), Puratronic™, 99.999% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
043822.KS1 Ea.
Catalog number 043822.KS
Price (USD)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7440-50-8
ChEBICHEBI:30052
Chemical Name or MaterialCopper plate
EINECS Number231-159-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only