2-Nitro-1-propanol, 97%
2-Nitro-1-propanol, 97%
2-Nitro-1-propanol, 97%
2-Nitro-1-propanol, 97%
Thermo Scientific Chemicals

2-Nitro-1-propanol, 97%

CAS: 2902-96-7 | C3H7NO3 | 105.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 043843.06
also known as 043843-06
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.00
Save 9.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.00
Save 9.35 (15%)
Each
Add to cart
Specifications
CAS2902-96-7
Chemical Name or Material2-Nitro-1-propanol
Density1.185 g/mL
Flash Point100°C (212°F)
Formula Weight105.09
View more
2-Nitro-1-propanol is used in improved synthesis of 2,2,2-trinitroethyl ethers, inhibits methane production during in vitro ruminal fermentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only