Ninhydrin, ACS reagent
Ninhydrin, ACS reagent
Ninhydrin, ACS reagent
Thermo Scientific Chemicals

Ninhydrin, ACS reagent

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 043846.22
Price (USD)
199.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
199.00
Each
Chemical Identifiers
CAS485-47-2
IUPAC Name2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
Molecular FormulaC9H6O4
InChI KeyFEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILESOC1(O)C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals
Impurity contentIdentification and melting point P.T.
Appearance (Color)White to brownish-white
CommentSensitivity to amino acids P.T.
Solution TestP.T.
Reagent for determination of amino acids and amine containing compounds, as an intermediate.Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Reagent for determination of amino acids and amine containing compounds, as an intermediate
RUO – Research Use Only