Aluminum oxide, catalyst support, high surface area
Aluminum oxide, catalyst support, high surface area
Aluminum oxide, catalyst support, high surface area
Aluminum oxide, catalyst support, high surface area
Thermo Scientific Chemicals

Aluminum oxide, catalyst support, high surface area

High surface area | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 043855.A7
also known as 043855-A7
Price (USD)
757.65
Online Exclusive
842.00
Save 84.35 (10%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
757.65
Online Exclusive
842.00
Save 84.35 (10%)
Each
Specifications
CAS1344-28-1
Chemical Name or MaterialAluminum oxide
EINECS Number215-691-6
IUPAC Namedialuminium(3+) trioxidandiide
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
View more
Aluminum oxide finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only