Aluminum oxide, catalyst support, intermediate surface area
Aluminum oxide, catalyst support, intermediate surface area
Aluminum oxide, catalyst support, intermediate surface area
Aluminum oxide, catalyst support, intermediate surface area
Thermo Scientific Chemicals

Aluminum oxide, catalyst support, intermediate surface area

Form: S.A. 12m2/g, contains 17.9% SiO2 | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 043856.A1
also known as 043856-A1
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Add to cart
Specifications
AppearanceWhite pellets
CAS1344-28-1
Chemical Name or MaterialAluminum Oxide
ColorWhite
EINECS Number215-691-6
View more
Aluminum oxide finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only