Silicon, Oil based standard solution, Specpure™ Si 5000μg/g
Silicon, Oil based standard solution, Specpure™ Si 5000μg/g
Silicon, Oil based standard solution, Specpure™ Si 5000μg/g
Thermo Scientific Chemicals

Silicon, Oil based standard solution, Specpure™ Si 5000μg/g

 
Have Questions?
Catalog NumberQuantity
043867.18
also known as 043867-18
50 g
Catalog number 043867.18
also known as 043867-18
Price (USD)
87.65
Online Exclusive
97.30
Save 9.65 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
87.65
Online Exclusive
97.30
Save 9.65 (10%)
Each
Specifications
Chemical Name or MaterialSilicon, Oil based standard solution
MDL NumberMFCD00085311
Name NoteSi 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only