Silver, Oil based standard solution, Specpure™ Ag 5000μg/g
Silver, Oil based standard solution, Specpure™ Ag 5000μg/g
Silver, Oil based standard solution, Specpure™ Ag 5000μg/g
Thermo Scientific Chemicals

Silver, Oil based standard solution, Specpure™ Ag 5000μg/g

 
Have Questions?
Catalog NumberQuantity
043868.18
also known as 043868-18
50 g
Catalog number 043868.18
also known as 043868-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver, Oil based standard solution
MDL NumberMFCD00003397
Name NoteAg 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only