Sulfur, Oil based standard solution, Specpure™ S 5000μg/g
Sulfur, Oil based standard solution, Specpure™ S 5000μg/g
Sulfur, Oil based standard solution, Specpure™ S 5000μg/g
Thermo Scientific Chemicals

Sulfur, Oil based standard solution, Specpure™ S 5000μg/g

 
Have Questions?
Catalog NumberQuantity
043870.18
also known as 043870-18
50 g
Catalog number 043870.18
also known as 043870-18
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.20
Save 13.55 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.20
Save 13.55 (15%)
Each
Specifications
Chemical Name or MaterialSulfur, Oil based standard solution
MDL NumberMFCD00242296
Name NoteS 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only