Silicon carbide, beta-phase, nanopowder, 95% min
Silicon carbide, beta-phase, nanopowder, 95% min
Silicon carbide, beta-phase, nanopowder, 95% min
Thermo Scientific Chemicals

Silicon carbide, beta-phase, nanopowder, 95% min

CAS: 409-21-2 | CSi | 40.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 043884.22
also known as 043884-22
Price (USD)
992.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,168.00
Save 175.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
992.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,168.00
Save 175.35 (15%)
Each
Chemical Identifiers
CAS409-21-2
IUPAC Namemethanidylidynesilylium
Molecular FormulaCSi
InChI KeyHBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES[C-]#[Si+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95% min
Silicon carbide is used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Used in electronic applications, measure gas temperatures in an optical technique called thin filament pyrometry. Used in jewelry, steel production, grapheme production. It is also used in production of exponential resistors, wire sawing, construction industry, a filler in plastics and composites, ceramic sintered parts, ferrites and soft metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only