Zinc 2-methoxyethoxide, 5% w/v in 2-methoxyethanol
Zinc 2-methoxyethoxide, 5% w/v in 2-methoxyethanol
Zinc 2-methoxyethoxide, 5% w/v in 2-methoxyethanol
Thermo Scientific Chemicals

Zinc 2-methoxyethoxide, 5% w/v in 2-methoxyethanol

CAS: 129918-15-6 | C6H16O4Zn | 217.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 043903.AE
also known as 043903-AE
Price (USD)
412.65
Special offer
Online exclusive
Ends: 30-Jun-2026
485.00
Save 72.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
412.65
Special offer
Online exclusive
Ends: 30-Jun-2026
485.00
Save 72.35 (15%)
Each
Specifications
CAS129918-15-6
Chemical Name or MaterialZinc 2-methoxyethoxide
ColorYellow
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
Density0.96 g/mL
View more
Zinc 2-methoxyethoxide is used as catalyst component in oxirane polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only