(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers
(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers
(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers
Thermo Scientific Chemicals

(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers

As a crosslinking agent | CAS: 74113-77-2 | H2NC2H4NHCH2C6H4C2H4Si(OCH3)3
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 043904.09
also known as 043904-09
Price (USD)
113.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
113.00
Each
Chemical Identifiers
CAS74113-77-2
IUPAC Name3,3-bis(2,2,6,6-tetramethylpiperidin-4-yl) 1,5-dioxospiro[5.5]undeca-7,9-diene-3,3-dicarboxylate
Molecular FormulaC31H46N2O6
InChI KeyIIZYNJMWYJKDJN-UHFFFAOYSA-N
SMILESCC1(C)CC(CC(C)(C)N1)OC(=O)C1(CC(=O)C2(CC=CC=C2)C(=O)C1)C(=O)OC1CC(C)(C)NC(C)(C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale Yellow
FormLiquid
Assay from Suppliers CofA≥88% (all isomers)
As a crosslinking agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only